Learn More
STEMCELL Technologies Mouse Pancreatic Organoids, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 70933
Encompass
Cryopreserved Mouse Pancreatic Organoids provide a convenient way to establish or standardize pancreatic organoid cultures in the laboratory. Each vial contains fragments of mouse pancreatic exocrine organoids sufficient to establish two wells of organoid cultures. The organoids are derived from the pancreas of C57BL/6 mice, cultured in PancreaCult™ Organoid Growth Medium (Mouse; Catalog #06040) and cryopreserved in CryoStor CS10 (Catalog #07930). Using cryopreserved Mouse Pancreatic Organoids enables establishment of pancreatic organoid cultures without the need to isolate pancreatic ducts from primary tissue, eliminating the need for access to fresh mouse tissue. The organoids can be passaged and expanded using PancreaCult™ Organoid Growth Medium (Mouse) and cryopreserved in CryoStor CS10. Mouse pancreatic exocrine organoid cultures can be used for research in a variety of fields, including epithelial cell biology, cancer, and metabolism.
Catalog No. NC1471926
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov