Learn More
Biomatik Corporation Mouse Paraoxonase 2 (PON2) ELISA Kit
Supplier:  Biomatik Corporation EKU06478
Encompass_Preferred
Mouse Paraoxonase 2 (PON2) ELISA Kit for detecting PON2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.052ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62086. Gene ID: 330260. Target Synonyms: Serum Paraoxonase/Arylesterase 2; Paraoxonase Nirs; Aromatic esterase 2; Serum aryldialkylphosphatase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5352