Learn More
American Research Products Inc Mouse parathyroid hormone PTH
Catalog No. 5000329613
Encompass
Mouse parathyroid hormone (PTH) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum tissue homogenates Method Sandwich
Catalog No. 50-003-29613 Supplier American Research Products Inc Supplier No. CSBE08690M
Mouse parathyroid hormone (PTH) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum tissue homogenates Method Sandwich