Learn More
BIOHIPPO Mouse PAX6 Microsample
SDP
Catalog No. 5000315210
Encompass
Mouse PAX6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PAX6 in Tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 25 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PAX6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PAX6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PAX6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15210 Supplier BIOHIPPO Supplier No. BHE15210695
Mouse PAX6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PAX6 in Tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 25 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PAX6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PAX6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PAX6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color