Learn More
Biomatik Corporation Mouse Paxillin (PXN) ELISA Kit
Supplier:  Biomatik Corporation EKU09091
Encompass_Preferred
Mouse Paxillin (PXN) ELISA Kit for detecting PXN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VI36. Gene ID: 19303. Target Synonyms: Pax. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8568