Learn More
BIOHIPPO Mouse PCDH15 Microsample
SDP
Catalog No. 5000313755
Encompass
Mouse PCDH15 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCDH15 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PCDH15 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PCDH15 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PCDH15 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13755 Supplier BIOHIPPO Supplier No. BHE15211200
Mouse PCDH15 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCDH15 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PCDH15 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PCDH15 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PCDH15 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color