Learn More
Proteintech Group Inc Mouse PCSK9 ELISA Kit
SDP
Supplier:  Proteintech Group Inc KE1005096T
Encompass_Preferred
KE10050 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The PCSK9 ELISA kit is to be used to detect and quantify protein levels of endogenous PCSK9. The assay recognizes mouse PCSK9. An antibody specific for PCSK9 has been pre-coated onto the microwells. The PCSK9 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for PCSK9 is added to detect the captured PCSK9 protein. For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Catalog No. 50-225-8457