Learn More
Sino Biological Mouse PCSK9 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785720
Encompass_Preferred
The Mouse PCSK9 ELISA Pair Set is for the quantitative determination of Mouse PCSK9.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for PCSK9 coated on a 96-well plate. Standards and samples are added to the wells, and any PCSK9 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-PCSK9 polyclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of PCSK9 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85720 Supplier Sino Biological Supplier No. SEK502515
The Mouse PCSK9 ELISA Pair Set is for the quantitative determination of Mouse PCSK9.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for PCSK9 coated on a 96-well plate. Standards and samples are added to the wells, and any PCSK9 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-PCSK9 polyclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of PCSK9 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.