Learn More
Sino Biological Anti-PCSK9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50251T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PCSK9 (rM PCSK9; Catalog#50251-M08H; NP_705793.1; Met1-Gln694). PCSK9 specific IgG was purified by Mouse PCSK9 affinity chromatography.
Catalog No. 50-241-1653