Learn More
Biomatik Corporation Mouse PCSK9 (Proprotein convertase subtilisin/kexin type 9) ELISA Kit
Supplier:  Biomatik Corporation EKF58650
Encompass_Preferred
Mouse PCSK9 (Proprotein convertase subtilisin/kexin type 9) ELISA Kit for detecting PCSK9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q80W65. Target Synonyms: PCSK9; PCSK9; Proprotein Convertase Subtilisin; Kexin Type 9; Proprotein Convertase 9; FH3; HCHOLA3; NARC-1; PC9; HCHOLA3; LDLCQ1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5441