Learn More
ACROBiosystems Mouse PD-1 100ug
Catalog No. 502534315
Encompass_Preferred
Mouse PD-1 Protein Mouse IgG2a Fc Tag (PD1-M5257) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q02242-1).
Catalog No. 50-253-4315 Supplier ACROBiosystems Supplier No. PD1M5257100UG
Mouse PD-1 Protein Mouse IgG2a Fc Tag (PD1-M5257) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q02242-1).