Learn More
BPS Bioscience Inc Mouse PD-1 PD-L1 Inhibitor Kit, ELISA assay, 96-well plate format, 100 binding reactions, chemiluminescence detection, biotinylated PD-1 and purified PD-L1
SDP
Supplier:  BPS Bioscience Inc 72027
Encompass
Mouse PD-1 PD-L1 Inhibitor Kit, ELISA assay, 96-well plate format, 100 binding reactions, chemiluminescence detection, biotinylated PD-1 and purified PD-L1
Catalog No. NC3357719