Learn More
ACROBiosystems Mouse PD-L1 25ug
Catalog No. 502534308
Encompass_Preferred
Biotinylated Mouse PD-L1 Protein HisAvitag (PDL-M82E7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # Q9EP73).
Catalog No. 50-253-4308 Supplier ACROBiosystems Supplier No. PDLM82E725UG
Biotinylated Mouse PD-L1 Protein HisAvitag (PDL-M82E7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # Q9EP73).