Learn More
Sino Biological Recombinant Anti-PD-1 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 50124R010H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PD1 / PDCD1 / CD279 (Catalog#50124-M08H; NP_032824.1; Met1-Gln167)and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1134