Learn More
Sino Biological Recombinant Anti-PD-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50124R345100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PD-1 / PDCD1 (rM PD-1 / PDCD1; Catalog#50124-M08H; NP_032824.1; Met1-Gln167).
Catalog No. 50-240-9835