Learn More
Sino Biological Recombinant Mouse PD-1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50124M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 167) of mouse PDCD1 (NP_032824.1) precursor was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3452