Learn More
Sino Biological Recombinant Anti-PD-L2/B7-DC Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50804R208100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PD-L2/B7-DC/CD273 (rM PD-L2/B7-DC/CD273; Catalog#50804-M08H; Q9WUL5; Met1-Arg219).
Catalog No. 50-241-0608