Learn More
Sino Biological Anti-PD-L2/B7-DC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50804RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse B7-DC / PD-L2 / CD273 (Catalog#50804-M08H; Q9WUL5; Met1-Arg219). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8531