Learn More
Sino Biological Recombinant Mouse PD-L2/B7-DC Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50804M08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse PDCD1LG2 (Q9WUL5) (Met1-Arg219) was expressed, with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3072