Learn More
BIOHIPPO Mouse PDE1C Microsample
SDP
Catalog No. 5000315650
Encompass
Mouse PDE1C Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PDE1C in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PDE1C Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PDE1C Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PDE1C biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15650 Supplier BIOHIPPO Supplier No. BHE15210892
Mouse PDE1C Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PDE1C in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PDE1C Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PDE1C Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PDE1C biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color