Learn More
FUJIFILM BIOSCIENCES INC Mouse PDGF-AA, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200541MG
Encompass_Preferred
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis.  PDGF synthesis is induced by IL-1, IL-6, TNF-A, TGF-B, and EGF signaling.  PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.  PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues.  PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB).  The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-A and PDGFR-B. 
Catalog No. 50-900-534