Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse PDGF-AA Recombinant Protein

Catalog No. 5018567
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-67 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-185-67 Supplier Invitrogen™ Supplier No. 14898980
Add to Cart
Add to Cart

Recombinant Protein

Description: The platelet-derived growth factor AA (PDGF-AA) is a member of the PDGF family. The PDGF proteins are derived from four genes (PDGF-A, -B, -C, and -D) that form four disulfide-linked homodimers (PDGF-AA, -BB, -CC, and -DD) and one heterodimer (PDGF-AB). PDGF-AA plays an important role as a mitogen for a number of cell types. Recombinant Mouse PDGF-AA produced in E.coli is a non-glycosylated homodimer of 126 amino acids with a molecular weight of 28.9 kDa. Applications Reported: Recombinant mouse PDGF-AA is biologically active and can promote proliferation of mouse 3T3 cells in culture. Applications Tested: This reagent has been tested in bioassays using the mouse cell line 3T3. The ED50 measured in a 3T3 proliferation assay is typically 10 ng/mL, corresponding to a specific activity of approximately 1 x10e5 Units/mg. Bioactivity: The ED50 measured in a 3T3 proliferation assay is typically 9.7 ng/mL, corresponding to a specific activity of approximately 1 x10e5 Units/mg. Endotoxin: Less than 0.1 ng/μg cytokine as determined by the LAL assay. Purity: >97% as determined by SDS-PAGE. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Platelet-derived growth factor (PDGF) was discovered as a major mitogenic factor present in serum but absent from plasma. It was found to be secreted from the alpha -granules of platelets activated during the coagulation of blood to form serum. Subsequent studies have demonstrated that PDGF is not one molecule but three, each a dimeric combination of two distinct but structurally related peptide chains designated A and B. The dimeric isoforms PDGF-AA, AB and BB are differentially expressed in various cell types and their effects are mediated through two distinct receptors, termed alpha and beta. Differences exist in isoform binding to each receptor. In general, PDGF isoforms are potent mitogens for connective tissue cells, including dermal fibroblasts, glial cells, arterial smooth muscle cells and some epithelial and endothelial cells. In addition to its activity as a mitogen, PDGF is chemotactic for fibroblasts, smooth muscle cells, neutrophils and mononuclear cells. Other reported activities for PDGF include stimulation of granule release by neutrophils and monocytes, facilitation of steroid synthesis by Leydig cells, stimulation of neutrophil phagocytosis, inhibition of natural killer (NK) cell activity, stimulation of collagen synthesis, modulation of thrombospondin expression and secretion, stimulation of collagenase activity and secretion, induction of contraction of rat aorta strips in vitro, and transient induction of T cell IL-2 secretion accompanied by a down-regulation of IL-4 and IFN-gamma production, temporary effects that may allow clonal expansion of antigen-activated B and T helper lymphocytes prior to differentiation. PDGF also appears to be ubiquitous in neurons throughout the CNS, where it is suggested to play an important role in neuron survival and regeneration, and in mediation of glial cell proliferation and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20033
Concentration 0.1 mg/mL
For Use With (Application) Bioactivity, Functional Assay
Formulation PBS with no preservative; pH 7.2
Gene ID (Entrez) 18590
Molecular Weight (g/mol) 28.9 kDa
Name Mouse PDGF-AA
pH Range 7.2
Purification Method Purified
Quantity 25 μg
Source E. coli
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration ≤0.1 ng/μg
Gene Alias H-PDGF-AA; PDGF A-chain; PDGF subunit A; PDGF1; PDGF-1; Pdgfa; PDGF-A; PDGFACP; platelet derived growth factor subunit A; platelet derived growth factor, alpha; Platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor subunit A; Rpa1
Common Name PDGF-AA
Gene Symbol Pdgfa
Protein Length 126 aa
Biological Activity ED50 = 9.7 ng/mL; determined by the dose-dependent proliferation of mouse 3T3 cells.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence 126aa
Content And Storage -80°C
Activity ED50 = 9.7 ng/mL; determined by the dose-dependent proliferation of mouse 3T3 cells.
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.