Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse PDGF-AB ELISA Kit

Catalog No. EM62RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM62RBX5 5 x 96 Tests
EM62RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EM62RBX5 Supplier Invitrogen™ Supplier No. EM62RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Mouse PDGF-AB quantitates mouse PDGF-AB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse PDGF-AB.

Platelet-Derived Growth Factor (PDGF) is a mitogenic peptide growth hormone carried in the alpha-granules of platelets and is released when platelets adhere to traumatized tissues. Connective tissue cells near the traumatized region respond by initiating the process of replication. The synthesis of PDGF can be induced by IL-1, IL-6, TNF-alpha, TGF-beta and EGF.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20033, P31240
Assay Range 26-6000 pg/mL
Assay Sensitivity 26 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Becaplermin; c-sis; H-PDGF-AB; IBGC5; PDGF A-chain; PDGF subunit A; PDGF subunit B; PDGF, B chain; PDGF1; PDGF-1; PDGF2; PDGF-2; Pdgfa; PDGF-A; PDGFAB; PDGFB; PDGF-B; platelet derived growth factor subunit A; platelet derived growth factor subunit B; platelet derived growth factor, alpha; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor B chain; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit A; Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); proto-oncogene c-Sis; SIS; SSVBecaplermin; c-sis; H-PDGF-AB; IBGC5; PDGF A-chain; PDGF subunit A; PDGF subunit B; PDGF, B chain; PDGF1; PDGF-1; PDGF2; PDGF-2; Pdgfa; PDGF-A; PDGFAB; PDGFB; PDGF-B; platelet derived growth factor subunit A; platelet derived growth factor subunit B; platelet derived growth factor, alpha; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor B chain; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit A; Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); proto-oncogene c-Sis; SIS; SSV;
Gene ID (Entrez) 18590, 18591
Gene Symbol PDGFA, PDGFB
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PDGF-AB
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.