Learn More
Sino Biological Recombinant Mouse PDGF-B Protein (His Tag)
Supplier:  Sino Biological 50801M07Y1
Encompass_Preferred
A DNA sequence encoding the mouse PDGFB (AAH23427.1) (Ser82-Thr190) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3074