Learn More
Sino Biological Anti-PDGF-D Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106721T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse PDGF-D/PDGFD, and purified by antigen affinity chromatography.
Catalog No. 50-241-1532