Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse PD-L1 / B7-H1, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;PD1,PDL1,PD-L1,CD274,B7-H1,B7H1,B7-H,B7H,PDCD1L1,PDCD1LG1
Supplier:  ACROBiosystems PD1M82F525UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse PD-L1 / B7-H1, Fc Tag, Avi Tag (Avitag™);25UG;MABSol Biotinylated Mouse PD-L1 / B7-H1, Fc Tag (PD1-M82F5) is expressed from human HEK293 cells. It contains AA Phe 19 - Thr 238 (Accession # NP_068693).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 53.2 kDa. The protein migrates as 65-85 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Mouse PD-1, Fc Tag (Cat. No. PD1-M5259) at 10 g/mL (100 l/well),can bind Biotinylated Mouse PD-L1, Fc Tag (Cat. No. PD1-M82F5) with a linear range of 0.15-2.5 g/mL (QC tested).
Catalog No. 50-108-9686