Learn More
Sino Biological Anti-Podoplanin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50256T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Podoplanin/PDPN (rM Podoplanin/PDPN; Catalog#50256-M08B1; NP_034459.2; Met1-Lys133). Podoplanin/PDPN specific IgG was purified by Mouse Podoplanin/PDPN affinity chromatography.
Catalog No. 50-241-1611