Learn More
Sino Biological Anti-Podoplanin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106682T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse Podoplanin, and purified by antigen affinity chromatography.
Catalog No. 50-241-1475