Learn More
Sino Biological Mouse PECAM1 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 50408R008P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD31/PECAM-1 (rM CD31/PECAM-1; Catalog#50408-M08H; NP_033868.3; Met1-Lys590) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-3353