Learn More
Biomatik Corporation Mouse Pepsin (PP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11238
Encompass_Preferred
Mouse Pepsin (PP) Mini sample ELISA Kit for detecting PP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.23-100ng/ml. Sensitivity: 0.51ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8189