Learn More
Biomatik Corporation Mouse Peptidase Inhibitor 16 (PI16) ELISA Kit
Supplier:  Biomatik Corporation EKU12361
Encompass_Preferred
Mouse Peptidase Inhibitor 16 (PI16) ELISA Kit for detecting PI16 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ET66. Gene ID: 74116. Target Synonyms: MSMBBP; CRISP9; PSPBP; Microseminoprotein, Beta-Binding Protein; Protease Inhibitor 16; PSP94-binding protein; Cysteine-rich secretory protein 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4911