Learn More
Biomatik Corporation Mouse Perforin 1 (PRF1) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10568
Encompass_Preferred
Mouse Perforin 1 (PRF1) Wide-range ELISA Kit for detecting PRF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10820. Gene ID: 18646. Target Synonyms: FLH2; HPLH2; P1; PFP; Pore Forming Protein; Cytolysin; Lymphocyte pore-forming protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7386