Learn More
Biomatik Corporation Mouse Perilipin 1 (PLIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU06531
Encompass_Preferred
Mouse Perilipin 1 (PLIN1) ELISA Kit for detecting PLIN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CGN5. Gene ID: 103968. Target Synonyms: PERI; PLIN1; Lipid Droplet Associated Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4638