Learn More
Biomatik Corporation Mouse Perilipin 4 (PLIN4) CLIA Kit
Supplier:  Biomatik Corporation EKU11647
Encompass_Preferred
Mouse Perilipin 4 (PLIN4) CLIA Kit for detecting PLIN4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.55pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O88492. Gene ID: 57435. Target Synonyms: Adipocyte protein S3-12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8107