Learn More
Biomatik Corporation Mouse Peroxiredoxin 4 (PRDX4) ELISA Kit
Supplier:  Biomatik Corporation EKU06555
Encompass_Preferred
Mouse Peroxiredoxin 4 (PRDX4) ELISA Kit for detecting PRDX4 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08807. Gene ID: 53381. Target Synonyms: Antioxidant enzyme AOE372; Thioredoxin-dependent peroxide reductase A0372. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6335