Learn More
Biomatik Corporation Mouse Peroxiredoxin 6 (PRDX6) ELISA Kit
Supplier:  Biomatik Corporation EKU06558
Encompass_Preferred
Mouse Peroxiredoxin 6 (PRDX6) ELISA Kit for detecting PRDX6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08709. Gene ID: 11758. Target Synonyms: 1-Cys; AOP2; NSGPx; PRX; aiPLA2; p29; Acidic calcium-independent phospholipase A2; Antioxidant protein 2; Non-selenium glutathione peroxidase; Red blood cells page spot 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7270