Learn More
Biomatik Corporation Mouse Peroxisome Proliferator Activated Receptor Delta (PPARd) ELISA Kit
Supplier:  Biomatik Corporation EKU06563
Encompass_Preferred
Mouse Peroxisome Proliferator Activated Receptor Delta (PPARd) ELISA Kit for detecting PPARd in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35396. Gene ID: 19015. Target Synonyms: FAAR; NR1C2; NUC1; NUCI; PPARB; PPARd/B; Nuclear Receptor Subfamily 1,Group C,Member 2; Nuclear hormone receptor 1; Peroxisome proliferator-activated receptor beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4356