Learn More
Raybiotech Inc MOUSE PF-4 ELISA
Supplier:  Raybiotech Inc ELMPF41
Encompass_Preferred
RayBio Mouse Platelet Factor 4 (CXCL4) ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 6 pg/ml - 3000 pg/ml
Catalog No. 50-104-4773