Learn More
Sino Biological Recombinant Mouse CXCL4/PF4 Protein (His Tag)
Catalog No. 502993155
Encompass_Preferred
A DNA sequence encoding the Mouse PF4 (Q6P8R3) (Val30-Ser105) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3155 Supplier Sino Biological Supplier No. 5A7692M07E1
A DNA sequence encoding the Mouse PF4 (Q6P8R3) (Val30-Ser105) was expressed, with a polyhistidine tag at the N-terminus.