Learn More
Sino Biological Mouse PGF Protein 500ug (Sino Biological)
Supplier:  Sino Biological 50125MCCH500
Encompass_Preferred
A DNA sequence encoding the mouse PGF (P49764) (Met1-Pro158) was fused with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-237-3802