Learn More
Sino Biological Mouse PGF Protein 50ug (Sino Biological)
Supplier:  Sino Biological 50125M02H50
Encompass_Preferred
A DNA sequence encoding the mouse PGF (P49764) (Met 1-Pro 158) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1642