Learn More
American Research Products Inc Mouse phosphoenolpyruVate carb
Catalog No. 5000327457
Encompass
Mouse phosphoenolpyruVate carboXykinase PCK ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27457 Supplier American Research Products Inc Supplier No. CSBE13613M
Mouse phosphoenolpyruVate carboXykinase PCK ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich