Learn More
Biomatik Corporation Mouse Phosphoglycerate Kinase 1 (PGK1) ELISA Kit
Supplier:  Biomatik Corporation EKU12414
Encompass_Preferred
Mouse Phosphoglycerate Kinase 1 (PGK1) ELISA Kit for detecting PGK1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09411. Gene ID: 18655. Target Synonyms: MIG10; PGKA; PRP 2; Cell migration-inducing gene 10 protein; Primer recognition protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6912