Learn More
Biomatik Corporation Mouse Phosphoinositide-3-Kinase Class-2-Alpha Polypeptide (PIK3C2a) ELISA Kit
Supplier:  Biomatik Corporation EKU06607
Encompass_Preferred
Mouse Phosphoinositide-3-Kinase Class-2-Alpha Polypeptide (PIK3C2a) ELISA Kit for detecting PIK3C2a in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61194. Gene ID: 18704. Target Synonyms: PI3K2a; CPK; PI3-K-C2 (Alpha) ; PI3-K-C2A; PI3K2-a; Phosphoinositide 3-Kinase 2a; Phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4399