Learn More
Biomatik Corporation Mouse Phospholipid Transfer Protein (PLTP) ELISA Kit
Supplier:  Biomatik Corporation EKU06634
Encompass_Preferred
Mouse Phospholipid Transfer Protein (PLTP) ELISA Kit for detecting PLTP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55065. Gene ID: 18830. Target Synonyms: Lipid transfer protein II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6121