Learn More
Braintree Scientific Mouse Pie Cage for Aerosol Delivery
SDP
Catalog No. 501955138
Encompass
Modified Mouse Pie Cage to accept aerosol delivery. Aerosol enters the PIE via tubing attached to the central core. Core is open on the top and solid on the bottom. PIE rotates so mice can be placed in individual sections. Aerosol exits each PIE section through one 3mm hole in the center of the outer edge. Aerosol is distributed to each PIE section via five 3mm holes. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5138 Supplier Braintree Scientific Supplier No. MPC3AERO
Modified Mouse Pie Cage to accept aerosol delivery. Aerosol enters the PIE via tubing attached to the central core. Core is open on the top and solid on the bottom. PIE rotates so mice can be placed in individual sections. Aerosol exits each PIE section through one 3mm hole in the center of the outer edge. Aerosol is distributed to each PIE section via five 3mm holes. **Sold for research purposes only and not for human use**