Learn More
American Research Products Inc Mouse pigment epithelium deriV
Catalog No. 5000329644
Encompass
Mouse pigment epithelium-deriVed factor PEDF ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-29644 Supplier American Research Products Inc Supplier No. CSBE08820M
Mouse pigment epithelium-deriVed factor PEDF ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich