Learn More
Biomatik Corporation Mouse Pigment Epithelium Derived Factor (PEDF) CLIA Kit
Supplier:  Biomatik Corporation EKU09811
Encompass_Preferred
Mouse Pigment Epithelium Derived Factor (PEDF) CLIA Kit for detecting PEDF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P97298. Gene ID: 20317. Target Synonyms: SERPINF1; EPC1; PIG35; SDF3; Serpin F1; Cell proliferation-inducing gene 35; Serpin Peptidase Inhibitor,Clade F Member 1 (Alpha-2 Antiplasmin) ; Stromal Cell Derived Factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5229