Learn More
Sino Biological Mouse PIGR ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372702
Encompass_Preferred
Each vial contains 50 ng of recombinant mouse PIGR. Reconstitute standard powder with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.
Catalog No. 50-237-2702 Supplier Sino Biological Supplier No. SEK501195
Each vial contains 50 ng of recombinant mouse PIGR. Reconstitute standard powder with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.