Learn More
Sino Biological Recombinant Anti-pIgR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50119R04020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PIGR extracellular domain (rM PIGR; Catalog#50119-M08H; NP_035212.2; Met 1-Lys 645).
Catalog No. 50-240-8679